2-(Phenylthio)nicotinoylchlorid
Lieferant: Thermo Scientific
BTB08736.1
BTB08736.10
MAYBBTB08736.1EA
0
EUR
MAYBBTB08736.1
MAYBBTB08736.10
2-(Phenylthio)nicotinoylchlorid
2-(Phenylthio)nicotinoylchlorid
Formel:
C₁₂H₈ClNOS |
CAS-Nummer:
165249-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...