Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl) ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Gly-OBzl·HCl, H-DL-Gly-OBzl hydrochloride, Benzyl glycinate hydrochloride, H-DL-Gly-OBzl.HCl

H50247.03 H50247.06
H50247.03EA 38.6 EUR
H50247.03 H50247.06
Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl) ≥98%
Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl)

Purity: 98% CAS No.: 2462-31-9 Molecular Formula: C9H11NO2.HCl Formula Weight: 201.65 Storage Temperature: 2°C to 8°C Melting Point: 139-140° MDL No.: MFCD00035442

Formel: C₉H₁₁NO₂·HCl
Schmelzpunkt: 139…140 °C
MDL Number: MFCD00035442
CAS-Nummer: 2462-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR