Ammoniumcarbamat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Carbamic acid ammonium salt
M-Clarity™ quality level = MQ100
292834-100G
292834-25G
292834-500G
SIAL292834-100GEA
60.1
EUR
SIAL292834-100G
SIAL292834-25G
SIAL292834-500G
Ammoniumcarbamat, Sigma-Aldrich®
Ammoniumcarbamat
Formel:
H₂NCO₂NH₄ |
MDL Number:
MFCD00013010 CAS-Nummer: 1111-78-0 EINECS: 214-185-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...