Dimethylamin 2 M in THF
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H27665.AE
H27665.AU
H27665.AEEA
95.4
EUR
H27665.AE
H27665.AU
Dimethylamin 2 M in THF
Dimethylamin
CAS No.: 124-40-3
Molecular Formula: C2H7N
Formula Weight: 45.08
Flash Point: -36°(-33°F)
Boiling Point: ca 60°
Beilstein Reference: 605257
Merck Reference: 14,3228
UN No.: 2924
MDL No.: MFCD00008288
Formel:
C₂H₇N Siedepunkt: ∼60 °C Dichte: 0,85 Flammpunkt: –36 °C (–33 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008288 CAS-Nummer: 124-40-3 EINECS: 204-697-4 UN-Nummer: 2924 ADR: 3,II Merck Index: 14,3228 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...