1-(4-Bromphenyl)-2-pyrrolidinon ≥98%
Lieferant: Thermo Fisher Scientific
H50563.03
H50563.06
H50563.03EA
124
EUR
H50563.03
H50563.06
1-(4-Bromphenyl)-2-pyrrolidinon ≥98%
1-(4-Bromphenyl)-2-pyrrolidinon
Purity: 98%
CAS No.: 7661-32-7
Molecular Formula: C10H10BrNO
Formula Weight: 240.10
Boiling Point: 98-101°
MDL No.: MFCD00138502
Formel:
C₁₀H₁₀BrNO Siedepunkt: 98…101 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00138502 CAS-Nummer: 7661-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...