3-Brombenzo[b]thiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

494976-25ML 494976-5ML
SIAL494976-25MLEA 285 EUR
SIAL494976-25ML SIAL494976-5ML
3-Brombenzo[b]thiophen, Sigma-Aldrich®
3-Brombenzo[b]thiophen
Formel: C₈H₅BrS
MDL Number: MFCD00023009
CAS-Nummer: 7342-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR