3,5-Difluor-2-methoxyphenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (3,5-Difluoro-2-methoxyphenyl)boronic acid, 3,5-Difluoro-2-methoxybenzeneboronic acid

L19773.03 L19773.06 L19773.MD
L19773.03EA 70.4 EUR
L19773.03 L19773.06 L19773.MD
3,5-Difluor-2-methoxyphenylboronsäure ≥97%
3,5-Difluor-2-methoxyphenylboronsäure

Purity: 97% CAS No.: 737000-76-9 Molecular Formula: C7H7BF2O3 Formula Weight: 187.94 Melting Point: 112-115° MDL No.: MFCD03095353

Formel: C₇H₇BF₂O₃
Schmelzpunkt: 112…115 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03095353
CAS-Nummer: 737000-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR