Isochinolin-5-amin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Isoquinolin-5-amine, 5-Aminoisoquinoline

L01223.03 L01223.06 L01223.14
L01223.03EA 31.6 EUR
L01223.03 L01223.06 L01223.14
Isochinolin-5-amin 99%
Isochinolin-5-amin

Purity: 99% CAS No.: 1125-60-6 Molecular Formula: C9H8N2 Formula Weight: 144.18 Melting Point: 126-131° Beilstein Reference: 114465

Formel: C₉H₈N₂
Schmelzpunkt: 126…131 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006907
CAS-Nummer: 1125-60-6
EINECS: 214-408-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR