5-Brombenzo[b]thiophen
Lieferant: Thermo Scientific
Synonyme:
5-Bromobenzothiophene
CC24810DA
CC24810EA
MAYBCC24810DAEA
0
EUR
MAYBCC24810DA
MAYBCC24810EA
5-Brombenzo[b]thiophen
5-Brombenzo[b]thiophen
Formel:
C₈H₅BrS |
MDL Number:
MFCD03069318 CAS-Nummer: 4923-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...