(4-Hydroxyphenyl)acetonitril
Lieferant: Thermo Scientific
Synonyme:
4-Hydroxybenzyl cyanide, 2-(4-Hydroxyphenyl)acetonitrile
SB00735DA
SB00735EA
SB00735EE
MAYBSB00735DAEA
0
EUR
MAYBSB00735DA
MAYBSB00735EA
MAYBSB00735EE
(4-Hydroxyphenyl)acetonitril
(4-Hydroxyphenyl)acetonitril
Formel:
C₈H₇NO Siedepunkt: 330 °C (1013 hPa) Schmelzpunkt: 68…71 °C |
MDL Number:
MFCD00002383 CAS-Nummer: 14191-95-8 EINECS: 238-046-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...