(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-1,2,3,4-Tetrahydronaphthalen-1-amine
M-Clarity™ quality level = MQ100
668796-25G
668796-5G
SIAL668796-25GEA
206
EUR
SIAL668796-25G
SIAL668796-5G
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamin, Sigma-Aldrich®
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamin
Formel:
C₁₀H₁₃N |
MDL Number:
MFCD00671630 CAS-Nummer: 23357-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...