Phthaloyldichlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Phthaloyl chloride, o-Phthaloyl dichloride
79830-100ML
79830-500ML
SIAL79830-100MLEA
40
EUR
SIAL79830-100ML
SIAL79830-500ML
Phthaloyldichlorid, Sigma-Aldrich®
Phthaloyldichlorid
Formel:
C₈H₄Cl₂O₂ Molekulargewicht: 203.02 g/mol |
MDL Number:
MFCD00000666 CAS-Nummer: 88-95-9 EINECS: 201-869-0 UN-Nummer: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...