(2-Bromphenyl)aceton 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-(2-Bromophenyl)propan-2-one

350590010 350590050
ACRO350590010EA 13.9 EUR
ACRO350590010 ACRO350590050
(2-Bromphenyl)aceton 99%
(2-Bromphenyl)aceton
Formel: C₉H₉BrO
Molekulargewicht: 213.07 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03410431
CAS-Nummer: 21906-31-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5595 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR