Quinaldine red 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-(2-(4-Dimethylamino)styryl)-1-ethylquinolinium iodide, 2-(4-(Dimethylamino)styryl)-1-ethylquinolin-1-ium iodide
Purity: 95%
CAS No.: 117-92-0
Molecular Formula: C21H23IN2
Formula Weight: 430.33
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Green to black
Beilstein Reference: 3798055
Merck Reference: 14,8049
MDL No.: MFCD00011968
Formel:
C₂₁H₂₃IN₂ Schmelzpunkt: 240 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011968 CAS-Nummer: 117-92-0 EINECS: 204-221-5 Merck Index: 14,8049 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...