ω-Brom-2-fluoracetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Bromomethyl o-fluorophenyl ketone, 2-Bromo-2'-fluoroacetophenone, α-Bromo-2-fluoroacetophenone, 2-Fluorophenacyl bromide, 2-Bromo-1-(2-fluorophenyl)ethan-1-one, 2-Bromo-1-(2-fluorophenyl)ethanone

H34179.03 H34179.09
H34179.03EA 37.2 EUR
H34179.03 H34179.09
ω-Brom-2-fluoracetophenon ≥98%
ω-Brom-2-fluoracetophenon

Purity: 98% CAS No.: 655-15-2 Molecular Formula: C8H6BrFO Formula Weight: 217.04 UN No.: 3265 MDL No.: MFCD00278796

Formel: C₈H₆BrFO
Molekulargewicht: 217.04 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00278796
CAS-Nummer: 655-15-2
UN-Nummer: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR