3-Brom-o-toluylsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H55436.03
H55436.06
H55436.03EA
44.3
EUR
H55436.03
H55436.06
3-Brom-o-toluylsäure ≥97%
3-Brom-o-toluylsäure
Purity: 97%
CAS No.: 76006-33-2
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 152-156°
MDL No.: MFCD00270097
Formel:
C₈H₇BrO₂ Schmelzpunkt: 152…156 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00270097 CAS-Nummer: 76006-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...