Knochenentkalker, OSTEOMOLL® und OSTEOSOFT®
Lieferant: Merck
1.01728.1000 1.01736.1000 1.01728.9010 1.01736.2500
1.01728.1000EA 79.9 EUR
1.01728.1000 1.01736.1000 1.01728.9010 1.01736.2500
Knochenentkalker, OSTEOMOLL® und OSTEOSOFT®
Färbemittel Mikroskopie- und Färbereagenzien
Hard tissue like bones or teeth, and all specimens of calcified tissue like bone marrow punches require special pre-treatment before routine processing by a histological laboratory.
Osteomoll® decalcifies effectively and depending on the size and density of the tissue, can be used twice, provided that the solution is still clear after the first decalcification process.
Osteosoft® is used for gentle decalcification based on complex-forming agents, particularly if it is to be followed by immunohistological procedures.