Biopsy specimen processing, CellSafe™
Lieferant: CELLPATH
EBB-0102-10A EBE-0201-02A EBC-0102-01A EBE-0201-03A
720-2355EA 29.2 EUR
720-2355 720-2333 720-2332 720-2334
Biopsy specimen processing, CellSafe™
Einbettkassetten Biopsiekassetten
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
  • No tissue damage
  • Low reagent carry-over
The CellSafe™ fits in every embedding cassette.