3,5-Dichlor-1,2,4-thiadiazol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
376030010
376030050
376030250
ACRO376030010EA
49
EUR
ACRO376030010
ACRO376030050
ACRO376030250
3,5-Dichlor-1,2,4-thiadiazol 97%
3,5-Dichloro-1,2,4-thiadiazole
Formel:
C₂Cl₂N₂S Siedepunkt: 165…167 °C (760 mmHg) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00799286 CAS-Nummer: 2254-88-8 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5570 to 1.5600 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...