Tetrakis(dimethylamino)ethylen, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
674613-10G
674613-1G
674613-50G
SIAL674613-10GEA
386
EUR
SIAL674613-10G
SIAL674613-1G
SIAL674613-50G
Tetrakis(dimethylamino)ethylen, Sigma-Aldrich®
Tetrakis(dimethylamino)ethylen
Formel:
C₁₀H₂₄N₄ |
MDL Number:
MFCD00009601 CAS-Nummer: 996-70-3 EINECS: 213-638-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...