(4-Chlorphenylthio)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-((4-Chlorophenyl)thio)acetic acid

L08583.09 L08583.18
L08583.09EA 55.1 EUR
L08583.09 L08583.18
(4-Chlorphenylthio)essigsäure ≥98%
(4-Chlorphenylthio)essigsäure

Purity: 98% CAS No.: 3405-88-7 Molecular Formula: C8H7ClO2S Formula Weight: 202.66 Melting Point: 104-107° Beilstein Reference: 1871569

Formel: C₈H₇ClO₂S
Schmelzpunkt: 104…107 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00021762
CAS-Nummer: 3405-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR