α-Ketoglutarsäure Dinatriumsalz Hydrat, Supelco®
Lieferant: Merck
Synonyme:
Sodium 2-oxoglutarate dibasic hydrate, 2-Oxopentanedioic acid disodium salt hydrate
K3752-1KG
K3752-100G
K3752-5G
K3752-1KGEA
2020
EUR
K3752-1KG
K3752-100G
K3752-5G
α-Ketoglutarsäure Dinatriumsalz Hydrat, Supelco®
α-Ketoglutarsäure Dinatriumsalz Hydrat
Formel:
C₅H₄Na₂O₅·xH₂O |
MDL Number:
MFCD00150702 CAS-Nummer: 305-72-6 EINECS: 206-167-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...