2-Amino-4-hydroxy-6-methylpyrimidin 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-Amino-6-methylpyrimidin-4(1H)-one, 2-Amino-6-methyl-4-pyrimidinol
103830250
103831000
ACRO103830250EA
26.3
EUR
ACRO103830250
ACRO103831000
2-Amino-4-hydroxy-6-methylpyrimidin 99%
2-Amino-4-hydroxy-6-methylpyrimidine
Formel:
C₅H₇N₃O Molekulargewicht: 125.13 g/mol Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00006095 CAS-Nummer: 3977-29-5 EINECS: 223-612-1 |
Spezifikation Testergebnisse
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
Clarity of solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...