1-Methyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methylpyrazole-4-boronic acid pinacol ester, 1-Methyl-4-pyrazole boronic acid pinacol ester, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

M-Clarity™ quality level = MQ100

595314-1G 595314-25G 595314-5G
SIAL595314-1GEA 79.9 EUR
SIAL595314-1G SIAL595314-25G SIAL595314-5G
1-Methyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
1-Methyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₀H₁₇BN₂O₂
MDL Number: MFCD03789259
CAS-Nummer: 761446-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR