Melatonin zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

M-Clarity™ quality level = MQ200

8.14537.0001 8.14537.0005
8.14537.0001EA 70.1 EUR
8.14537.0001 8.14537.0005
Melatonin zur Synthese, Sigma-Aldrich®
Melatonin
Formel: C₁₃H₁₆N₂O₂
MDL Number: MFCD00005655
CAS-Nummer: 73-31-4
EINECS: 200-797-7

Bestellen

Spezifikation Testergebnisse

Melting Point 116 - 118 °C
Molar Mass 232.28 g/mol
Synonyms N-Acetyl-5-methoxytryptamine
Hill Formula C13H16N2O2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR