2,3-Dimethoxy-5-methyl-p-benzochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3-Dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione, 2,3-Dimethoxy-5-methyl-1,4-benzoquinone

D9150-1G D9150-5G
SIALD9150-1GEA 105 EUR
SIALD9150-1G SIALD9150-5G
2,3-Dimethoxy-5-methyl-p-benzochinon, Sigma-Aldrich®
2,3-Dimethoxy-5-methyl-p-benzochinon
Formel: C₉H₁₀O₄
MDL Number: MFCD00001595
CAS-Nummer: 605-94-7
EINECS: 210-100-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR