Methylmercaptoacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl thioglycolate

M-Clarity™ quality level = MQ200

108995-100G 108995-500G 108995-5G
SIAL108995-100GEA 42.5 EUR
SIAL108995-100G SIAL108995-500G SIAL108995-5G
Methylmercaptoacetat, Sigma-Aldrich®
Methylmercaptoacetat
Formel: C₃H₆O₂S
MDL Number: MFCD00004873
CAS-Nummer: 2365-48-2
EINECS: 219-121-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR