Hexylmercaptoacetat

Lieferant: Thermo Scientific

BTB11443.250 BTB11443.1
MAYBBTB11443.250EA 0 EUR
MAYBBTB11443.250 MAYBBTB11443.1
Hexylmercaptoacetat
Hexylmercaptoacetat
Formel: C₈H₁₆O₂S
CAS-Nummer: 20292-01-7
EINECS: 243-696-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR