2-Amino-4-ethylpyridin

Lieferant: Thermo Scientific

Synonyme: 4-Ethylpyridin-2-amine, 4-Ethyl-2-pyridylamine

BTB11175.1 BTB11175.10 BTB11175.50
MAYBBTB11175.1EA 0 EUR
MAYBBTB11175.1 MAYBBTB11175.10 MAYBBTB11175.50
2-Amino-4-ethylpyridin
2-Amino-4-ethylpyridine
Formel: C₇H₁₀N₂
MDL Number: MFCD00129025
CAS-Nummer: 33252-32-3
EINECS: 251-430-2
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR