(±)-Methyl-2-brompropionat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-bromopropanoate

M-Clarity™ quality level = MQ100

167185-100G 167185-25G
SIAL167185-100GEA 69.6 EUR
SIAL167185-100G SIAL167185-25G
(±)-Methyl-2-brompropionat, Sigma-Aldrich®
(±)-Methyl-2-brompropionat
Formel: C₄H₇BrO₂
MDL Number: MFCD00000143
CAS-Nummer: 5445-17-0
EINECS: 226-642-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR