Isopentylpropionat, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Isoamyl Propionate, Propionic acid isoamyl ester

M-Clarity™ quality level = MQ300

W208213-1KG-K W208213-4KG-K
SIALW208213-1KG-KEA 412 EUR
SIALW208213-1KG-K SIALW208213-4KG-K
Isopentylpropionat, Sigma-Aldrich®
Isopentylpropionat
Formel: C₈H₁₆O₂
MDL Number: MFCD00048611
CAS-Nummer: 105-68-0
EINECS: 203-322-1
UN-Nummer: 3272
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR