Aluminiumtri-sec-butanolat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide

M-Clarity™ quality level = MQ100

201073-100G 201073-500G
SIAL201073-100GEA 59.4 EUR
SIAL201073-100G SIAL201073-500G
Aluminiumtri-sec-butanolat, Sigma-Aldrich®
Aluminiumtri-sec-butanolat
Formel: C₁₂H₂₇AlO₃
MDL Number: MFCD00009327
CAS-Nummer: 2269-22-9
EINECS: 218-871-2
UN-Nummer: 1992
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR