Aluminiumtri-sec-butanolat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide
A13044.36
A13044.0E
A13044.22
A13044.36EA
114
EUR
A13044.36
A13044.0E
A13044.22
Aluminiumtri-sec-butanolat 97%
Aluminiumtri-sec-butanolat
Purity: 97%
CAS No.: 2269-22-9
Molecular Formula: Al[OCH(CH3)CH2CH3]3
Formula Weight: 246.32
Flash Point: 27°(82°F)
Refractive Index: 1.4390
Odour: Alcohol-like
Boiling Point: 200-206°/30mm
Beilstein Reference: 3910908
UN No.: 1992
MDL No.: MFCD00009327
Formel:
C₁₂H₂₇AlO₃ Siedepunkt: 200…206 °C (30 mmHg) Dichte: 0.967 Flammpunkt: 27 °C (82 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00009327 CAS-Nummer: 2269-22-9 EINECS: 218-871-2 UN-Nummer: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...