Azetidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

281069-1G 281069-250MG
SIAL281069-1GEA 166 EUR
SIAL281069-1G SIAL281069-250MG
Azetidin, Sigma-Aldrich®
Azetidin
Formel: C₃H₇N
MDL Number: MFCD00005165
CAS-Nummer: 503-29-7
EINECS: 207-963-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR