1,4-Naphthalindicarbonsäure 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L20249.14
L20249.22
L20249.14EA
84.7
EUR
L20249.14
L20249.22
1,4-Naphthalindicarbonsäure 98+%
1,4-Naphthalindicarbonsäure
Purity: 98+%
CAS No.: 605-70-9
Molecular Formula: C12H8O4
Formula Weight: 216.19
Melting Point: 321-325°
Beilstein Reference: 2051255
Formel:
C₁₂H₈O₄ Schmelzpunkt: 321…325 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00014312 CAS-Nummer: 605-70-9 EINECS: 210-094-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...