Rheniumdisulfid ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Rhenium (IV) sulphide

89482.04 89482.09
89482.04EA 385 EUR
89482.04 89482.09
Rheniumdisulfid ≥99%
Rheniumdisulfid

Purity: 99% CAS No.: 12038-63-0 Molecular Formula: ReS2 Formula Weight: 250.33 Physical Form: Powder Density: 7.506 MDL No.: MFCD00016307

Formel: ReS₂
Dichte: 7.506
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00016307
CAS-Nummer: 12038-63-0
EINECS: 234-878-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR