Chrom(VI)oxid ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

12522.30 12522.A1 12522.D9
12522.30EA 50.9 EUR
12522.30 12522.A1 12522.D9
Chrom(VI)oxid ≥99%
Chrom(VI)oxid

Purity: 99% CAS No.: 1333-82-0 Molecular Formula: CrO3 Formula Weight: 99.99 Physical Form: Crystalline Density: 2.7 Appearance: red Melting Point: 196° Boiling Point: 250° dec. Merck Reference: 14,2235 UN No.: 1463 MDL No.: MFCD00010952

Formel: CrO₃
Siedepunkt: 250 °C (dec.)
Schmelzpunkt: 196 °C
Dichte: 2.7
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010952
CAS-Nummer: 1333-82-0
EINECS: 215-607-8
UN-Nummer: 1463
ADR: 5.1,II
REACH-Nummer: 01-2119458868-17
Merck Index: 14,2235

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR