Zinkselenid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Zinc (II) selenide
10999.09
10999.18
10999.30
10999.09EA
84.2
EUR
10999.09
10999.18
10999.30
Zinkselenid ≥99,999% (Metall-Basis)
Zinkselenid
Purity: 99.999%
CAS No.: 1315-09-9
Molecular Formula: ZnSe
Formula Weight: 144.33
Physical Form: Powder
Density: 5.42
Melting Point:
Formel:
SeZn Molekulargewicht: 144.35 g/mol Schmelzpunkt: >1100 °C Dichte: 5.42 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011296 CAS-Nummer: 1315-09-9 EINECS: 215-259-7 UN-Nummer: 3283 ADR: 6.1 III Merck Index: 14,10156 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...