Zinkoxid ≥99,9995% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Zinci oxidum, Zinc (II) oxide
12855.14
12855.22
12855.14EA
173
EUR
12855.14
12855.22
Zinkoxid ≥99,9995% (Metall-Basis), Puratronic®
Zinkoxid
Purity: 99.9995%
CAS No.: 1314-13-2
Molecular Formula: ZnO
Formula Weight: 81.37
Physical Form: Powder
Density: 5.606
Melting Point: 1975°
Merck Reference: 14,10147
UN No.: 3077
MDL No.: MFCD00011300
Formel:
ZnO Molekulargewicht: 81.39 g/mol Dichte: 5.606 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011300 CAS-Nummer: 1314-13-2 EINECS: 215-222-5 UN-Nummer: 3077 ADR: 9 III REACH-Nummer: 01-2119463881-32 Merck Index: 14,10147 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...