Lanthan(III)chlorid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Lanthanum chloride, Lanthanum trichloride
ampouled under argon
Purity: 99.99%
CAS No.: 10099-58-8
Molecular Formula: LaCl3
Formula Weight: 245.27
Physical Form: Granular, Ampouled under argon
Density: 3.842
Appearance: White
Odour: Odourless
Melting Point: 860°
Boiling Point: 1000°
Merck Reference: 14,5363
UN No.: 3077
MDL No.: MFCD00011068
Formel:
Cl₃La Siedepunkt: 1000 °C Schmelzpunkt: 860 °C Dichte: 3.842 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011068 CAS-Nummer: 10099-58-8 EINECS: 233-237-5 UN-Nummer: 3077 ADR: 9,III Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...