Diarsentriselenid ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Arsenic(III) selenide

12849.06 12849.14
12849.06EA 157 EUR
12849.06 12849.14
Diarsentriselenid ≥99,999% (Metall-Basis)
Diarsentriselenid

Purity: 99.999% CAS No.: 1303-36-2 Molecular Formula: As2Se3 Formula Weight: 386.72 Physical Form: 1-6mm Fused Pieces Density: 4.75 Melting Point: 260° Merck Reference: 14,805 UN No.: 3077 MDL No.: MFCD00014166


For vacuum deposition

Formel: As₂Se₃
Schmelzpunkt: 260 °C
Dichte: 4.75
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00014166
CAS-Nummer: 1303-36-2
EINECS: 215-119-5
UN-Nummer: 3077
ADR: 9,III
Merck Index: 14,805

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR