di-Phosphorpentoxid 98%, reinst
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride
200890010
200890025
200890100
200892500
ACRO200890010EA
74.9
EUR
ACRO200890010
ACRO200890025
ACRO200890100
ACRO200892500
di-Phosphorpentoxid 98%, reinst
di-Phosphorpentoxid
Formel:
P₂O₅ Molekulargewicht: 141.94 g/mol Schmelzpunkt: 340…360 °C (Subl.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011440 CAS-Nummer: 1314-56-3 EINECS: 215-236-1 UN-Nummer: 1807 ADR: 8 II Merck Index: 13,07441 |
Spezifikation Testergebnisse
Appearance | White Hygroscopic crystalline powder |
Titration with NaOH | ≥97.5 % |
Reducing substance (P2O3) | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...