Antimon(III)chlorid analytisches Reagens, Supelco®
Lieferant: Merck
1.07838.0250
1.07838.1000
1.07838.0250EA
172
EUR
1.07838.0250
1.07838.1000
Antimon(III)chlorid analytisches Reagens, Supelco®
Antimon(III)chlorid
Spezifikation Testergebnisse
Solubility | 931 g/l (20 °C) Hydrolysis |
Melting Point | 73 °C |
Molar Mass | 228.1 g/mol |
Bulk Density | 1600 kg/m3 |
Boiling Point | 233 °C (1013 hPa) |
Vapor Pressure | 0.16 hPa (20 °C) |
Density | 3.14 g/cm3 (20 °C) |
pH | (H2O) acidic |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...