Kieselgel, Porengröße: 60 Å 0.032-0.063 mm (230-450 mesh)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L14003.36
L14003.0B
L14003.0E
L14003.36EA
69.8
EUR
L14003.36
L14003.0B
L14003.0E
Kieselgel, Porengröße: 60 Å 0.032-0.063 mm (230-450 mesh)
Silica gel
CAS No.: 7631-86-9
Odour: Odourless
Merck Reference: 14,8493
MDL No.: MFCD00011232
Formel:
SiO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011232 CAS-Nummer: 63231-67-4 EINECS: 231-545-4 Merck Index: 14,8493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...