Blei(II)chlorid, ultra trocken ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Lead dichloride

ampouled under argon

35704.06 35704.14 35704.22
35704.06EA 200 EUR
35704.06 35704.14 35704.22
Blei(II)chlorid, ultra trocken ≥99,999% (Metall-Basis)
Blei(II)chlorid

Purity: 99.999% CAS No.: 7758-95-4 Molecular Formula: PbCl2 Formula Weight: 278.10 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.85 Melting Point: 501° Boiling Point: 951° Merck Reference: 14,5404 UN No.: 2291 MDL No.: MFCD00011157

Formel: Cl₂Pb
Siedepunkt: 951 °C
Schmelzpunkt: 501 °C
Dichte: 5.85
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011157
CAS-Nummer: 7758-95-4
EINECS: 231-845-5
UN-Nummer: 2291
ADR: 6.1,III
Merck Index: 14,5404

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR