Blei(II)chlorid 99%, Reagenzqualität

Lieferant: Thermo Fisher Scientific
Gefahr

12349.18 12349.36 12349.A3
12349.18EA 25.1 EUR
12349.18 12349.36 12349.A3
Blei(II)chlorid 99%, Reagenzqualität
Blei(II)chlorid

Purity: 99% CAS No.: 7758-95-4 Molecular Formula: PbCl2 Formula Weight: 278.10 Physical Form: Crystalline Powder Density: 5.85 Melting Point: 501° Boiling Point: 951° Solubility: Soluble in hot water, alkali hydroxides, NH4Cl solution. Insoluble in cold water and alcohol Merck Reference: 14,5404 UN No.: 2291 MDL No.: MFCD00011157


Analytical reagent, preparation of lead salts, as solder and flux

Formel: Cl₂Pb
Siedepunkt: 951 °C
Schmelzpunkt: 501 °C
Dichte: 5,85
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011157
CAS-Nummer: 7758-95-4
EINECS: 231-845-5
UN-Nummer: 2291
ADR: 6.1,III
Merck Index: 14,5404

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR