(±)-Nicotin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

N0267-100MG N0267-1G N0267-500MG
SIALN0267-100MGEA 202 EUR
SIALN0267-100MG SIALN0267-1G SIALN0267-500MG
(±)-Nicotin, Sigma-Aldrich®
(±)-Nicotin
Formel: C₁₀H₁₄N₂
MDL Number: MFCD00065370
CAS-Nummer: 22083-74-5
UN-Nummer: 1654
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR