N,N-Dimethyl-1,3-diaminopropan, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
3-Dimethylaminopropylamine, N,N-Dimethyltrimethylenediamine, N,N-Dimethyl-1,3-propanediamine
M-Clarity™ quality level = MQ200
D145009-1L
D145009-500ML
D145009-25ML
SIALD145009-1LEA
50.1
EUR
SIALD145009-1L
SIALD145009-500ML
SIALD145009-25ML
N,N-Dimethyl-1,3-diaminopropan, Sigma-Aldrich®
N,N-Dimethyl-1,3-diaminopropan
Formel:
C₅H₁₄N₂ |
MDL Number:
MFCD00008216 CAS-Nummer: 109-55-7 EINECS: 203-680-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...