Cysteaminiumchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Aminoethanethiol hydrochloride, Cysteamine Hydrochloride, 2-Mercaptoethylamine hydrochloride, 2-MEA, β-Mercaptoethylamine hydrochloride, Mercaptoethylamine-HCl, H-Cys-NH2·HCl
M-Clarity™ quality level = MQ200
30080-100G
30080-25G
SIAL30080-100GEA
202
EUR
SIAL30080-100G
SIAL30080-25G
Cysteaminiumchlorid, Sigma-Aldrich®
Cysteaminiumchlorid
Formel:
C₂H₇NS·HCl |
MDL Number:
MFCD00012904 CAS-Nummer: 156-57-0 EINECS: 205-858-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...