Tetraethylenglycoldimethylether, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,5,8,11,14-Pentaoxapentadecane
M-Clarity™ quality level = MQ200
172405-1KG
172405-250G
SIAL172405-1KGEA
179
EUR
SIAL172405-1KG
SIAL172405-250G
Tetraethylenglycoldimethylether, Sigma-Aldrich®
Tetraethylenglycoldimethylether
Formel:
C₁₀H₂₂O₅ |
MDL Number:
MFCD00008505 CAS-Nummer: 143-24-8 EINECS: 205-594-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...